missing translation for 'onlineSavingsMsg'
Learn More

TCI America™ Tissue-Clearing Reagent CUBIC-L

Catalog No. T3740500ML
Change view
Click to view available options
Capacity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity
T3740500ML 500 mL
T374025ML 25 mL
T3740100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T3740500ML Supplier TCI America™ Supplier No. T3740500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Used along with CUBIC-R+(M) (T3741) for tissue-clearing. Note: both products are required are for the clearing procedure.

  • The CUBIC technique enables cyclopedic imaging at a single-cell resolution following whole-body and whole-organ clearing.
  • CUBIC-L (T3740) is used for delipidation while CUBIC-R+(M) (T3741) is used for RI matching.

Note: Both T3740 and T3741 are required for the CUBIC clearing process.

Specifications

Capacity 500 mL
For Use With (Equipment) Light-sheet Fluorescent Microscopy (LSFM) or Confocal Laser-scanning Microscopy (CLSM) Device
Product Line CUBIC
For Use With (Application) Neuroscience, 3D pathology, oncology, histology, immunology, and other research applications
Form Liquid