Learn More
TCI America™ Tissue-Clearing Reagent CUBIC-L
Used along with CUBIC-R+(M) (T3741) for tissue-clearing. Note: both products are required are for the clearing procedure.
Supplier: TCI America™ T3740100ML
Description
- The CUBIC technique enables cyclopedic imaging at a single-cell resolution following whole-body and whole-organ clearing.
- CUBIC-L (T3740) is used for delipidation while CUBIC-R+(M) (T3741) is used for RI matching.
Note: Both T3740 and T3741 are required for the CUBIC clearing process.
Chemical Identifiers
| Light-sheet Fluorescent Microscopy (LSFM) or Confocal Laser-scanning Microscopy (CLSM) Device | |
| CUBIC | |
| Neuroscience, 3D pathology, oncology, histology, immunology, and other research applications | |
| Liquid |
Specifications
| 100 mL | |
| CUBIC | |
| Liquid |
| Light-sheet Fluorescent Microscopy (LSFM) or Confocal Laser-scanning Microscopy (CLSM) Device | |
| Neuroscience, 3D pathology, oncology, histology, immunology, and other research applications |