missing translation for 'onlineSavingsMsg'
Learn More

TCI America™ Tissue-Clearing Reagent CUBIC-HL

Catalog No. T378125ML
Change view
Click to view available options
Capacity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity
T378125ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T378125ML Supplier TCI America™ Supplier No. T378125ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CUBIC-HL is optimized for clearing highly fatty tissues. Note this does not preserve the fluorescent signal of fluorescent proteins.

  • The CUBIC technique enables cyclopedic imaging at a single-cell resolution following whole-body and whole-organ clearing.
  • CUBIC-HL is an optional addition to the CUBIC clearing protocol, and can be used to clear highly fatty tissue and quenching autofluourescence.

Note: Both T3741 and T3740 are required for the CUBIC clearing process.

Specifications

Capacity 25 mL
For Use With (Equipment) Light-sheet Fluorescent Microscopy (LSFM) or Confocal Laser-scanning Microscopy (CLSM) Device
Product Line CUBIC
For Use With (Application) Neuroscience, 3D pathology, oncology, histology, immunology, and other research applications
Form Liquid