missing translation for 'onlineSavingsMsg'
Learn More

Tiropramide Hydrochloride 98.0+%, TCI America™

Catalog No. p-7137509
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T29781G 1 g
T29785G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T29781G Supplier TCI America Supplier No. T29781G

Chemical Identifiers

CAS 53567-47-8
Molecular Formula C28H42ClN3O3
Molecular Weight (g/mol) 504.11
MDL Number MFCD01741847
InChI Key SVCQSEFEZWMYSA-UHFFFAOYNA-N
PubChem CID 134448
IUPAC Name 3-{4-[2-(diethylamino)ethoxy]phenyl}-2-(phenylformamido)-N,N-dipropylpropanamide hydrochloride
SMILES Cl.CCCN(CCC)C(=O)C(CC1=CC=C(OCCN(CC)CC)C=C1)NC(=O)C1=CC=CC=C1

Specifications

Melting Point 182°C
Color White
Quantity 1 g
Formula Weight 504.11
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Tiropramide Hydrochloride