Learn More
Gilson TIPACK Adapter For PIPETMAN™ M96
This adapter allows the use of PIPETMAN™ DIAMOND Tips TIPACKS with PIPETMAN™ M96
Supplier:  Gilson FH07006
Catalog No. FH07006G
Only null left
Explore available promotions
Description
Description
  • Designed for the 96-channel motorized pipette PIPETMAN M96
  • Allows PIPETMAN M96 to use packaged/racked pipette tips of the type PIPETMAN DIAMOND Tips sold as TIPACKs.
  • Adapter provides the mechanical interface between the M96 pipette tip-loading mechanism and the TIPACK format tips, so you can seamlessly load 96-channel tips from racks rather than using loose tips
Specifications
Specifications
Tip Transfer Tool Fitting
1 Ea.
0.15 kg
Aluminium
0.34 lb.
Product Suggestions
Product Suggestions