missing translation for 'onlineSavingsMsg'
Learn More

Gilson™ Tip Disposal Bin

Catalog No. 32000274
Change view
Click to view available options
Autoclavable:
Not Autoclavable
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Autoclavable
32000274 Not Autoclavable
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 32000274 Supplier Gilson™ Supplier No. 32000274

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Bin for collecting spent tips from PIPETMAX.

  • Bin for collecting spent tips from PIPETMAX.

Specifications

Capacity (English) 36.18 oz.
Color White
Sterility Non-sterile
Autoclavable Not Autoclavable
For Use With (Application) For collecting spent PIPETMAX tips.
For Use With (Equipment) PIPETMAX Tips
Material Plastic
Product Type Disposal Container