Learn More
Labcon North America Eclipse™ 1200uL Extended Length Pipet Tips for Sartorius Pipettors, in Racks, Sterile
Supplier: Labcon North America 10489650089
Tip, Pipet; Labcon; Eclipse™; Extended Length; Fits Sartorius 1200uL pipettors; Low mounting and ejection forces less than 3kg; Sterile; ATP, RNase/DNase and Endotoxin Free (non-pyrogenic); Polypropylene USP Class VI resin; Autoclavable; Validated Sterilization (ISO 11137) for use in Research and Medical labs; 96 tips/rack; 6 racks per pack; 8 packs per case; 88.9mm. H; Made in USA