missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Tioconazole, 98+%

Potent inhibitor of cytochrome-P450 | CAS: 65899-73-2 | C16H13Cl3N2OS

Supplier:  Thermo Scientific Chemicals J6045906

 View more versions of this product

Catalog No. AAJ6045906


Only null left

Description

Description

Potent inhibitor of cytochrome-P450

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

5 g
Potent inhibitor of cytochrome-P450
65899-73-2
387.71g/mol
Signal Transduction Reagent
White to Off-white
≥98%
C16H13Cl3N2OS
Thioconazole
Powder

Chemical Identifiers

65899-73-2
C16H13Cl3N2OS
Thioconazole
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 265-973-8

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only