missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Tioconazole, 98+%

Catalog No. AAJ6045903
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6045903 1 g
AAJ6045906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6045903 Supplier Thermo Scientific Chemicals Supplier No. J6045903

Potent inhibitor of cytochrome-P450 | CAS: 65899-73-2 | C16H13Cl3N2OS

Potent inhibitor of cytochrome-P450

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 65899-73-2
Molecular Formula C16H13Cl3N2OS
Synonym Thioconazole

Specifications

Quantity 1 g
Description White to Off-white
For Use With (Application) Potent inhibitor of cytochrome-P450
Formula Weight 387.71g/mol
Product Type Signal Transduction Reagent
Form Powder
Assay ≥98%
EINECSNumber 265-973-8
Recommended Storage Ambient temperatures

RUO – Research Use Only