missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tinidazole USP, Medisca™

Description
Tinidazole USP is an almost white or pale yellow, crystalline powder.
Specifications
Specifications
| Physical Form | Powder |
| CAS | 19387-91-8 |
| Quantity | 500 g |
| Grade | Not Available |