Learn More
Thermo Scientific™ Tinidazole
Chemical Identifiers
| CAS | 19387-91-8 |
|---|---|
| Molecular Formula | C8H13N3O4S |
| Molecular Weight (g/mol) | 247.27 |
| MDL Number | MFCD00057217 |
| InChI Key | HJLSLZFTEKNLFI-UHFFFAOYSA-N |
| IUPAC Name | 1-[2-(ethanesulfonyl)ethyl]-2-methyl-5-nitro-1H-imidazole |
| SMILES | CCS(=O)(=O)CCN1C(C)=NC=C1[N+]([O-])=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AC463510050
|
Thermo Scientific™
463510050 |
5 g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AC463510250
|
Thermo Scientific™
463510250 |
25 g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 19387-91-8 | |
| 247.27 | |
| HJLSLZFTEKNLFI-UHFFFAOYSA-N | |
| CCS(=O)(=O)CCN1C(C)=NC=C1[N+]([O-])=O |
| C8H13N3O4S | |
| MFCD00057217 | |
| 1-[2-(ethanesulfonyl)ethyl]-2-methyl-5-nitro-1H-imidazole |
Specifications
| 19387-91-8 | |
| MFCD00057217 | |
| Solubility aq. soln.: insoluble. Other solubilities: soluble in acetone, slightly soluble in methanol, very slightly soluble on ethanol | |
| CCS(=O)(=O)CCN1C(C)=NC=C1[N+]([O-])=O | |
| 247.27 | |
| Tinidazole |
| C8H13N3O4S | |
| 5 g | |
| HJLSLZFTEKNLFI-UHFFFAOYSA-N | |
| 1-[2-(ethanesulfonyl)ethyl]-2-methyl-5-nitro-1H-imidazole | |
| 247.27 |
Safety and Handling
GHS H Statement Harmful in contact with skin.
Harmful if inhaled.
Suspected of causing genetic defects.
Suspected of causing cancer.
GHS P Statement IF ON SKIN: Wash with plenty of soap and water.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
GHS Signal Word: Warning
EINECSNumber : 243-014-4
RUO â Research Use Only