missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Tinidazole

Catalog No. AC463510250
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC463510250 25 g
AC463510050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC463510250 Supplier Thermo Scientific Chemicals Supplier No. 463510250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 19387-91-8
Molecular Formula C8H13N3O4S
Molecular Weight (g/mol) 247.27
MDL Number MFCD00057217
InChI Key HJLSLZFTEKNLFI-UHFFFAOYSA-N
IUPAC Name 1-[2-(ethanesulfonyl)ethyl]-2-methyl-5-nitro-1H-imidazole
SMILES CCS(=O)(=O)CCN1C(C)=NC=C1[N+]([O-])=O

Specifications

Melting Point 124.0°C to 128.0°C
Quantity 25 g
Solubility Information Solubility aq. soln.: insoluble. Other solubilities: soluble in acetone, slightly soluble in methanol, very slightly soluble on ethanol
Formula Weight 247.27
Chemical Name or Material Tinidazole
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement Harmful in contact with skin.

Harmful if inhaled.

Suspected of causing genetic defects.

Suspected of causing cancer.

Precautionary Statement GHS P Statement IF ON SKIN: Wash with plenty of soap and water.

Call a POISON CENTER or doctor/physician if you feel unwell.

IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.

Wear protective gloves/protective clothing/eye protection/face protection.

EINECSNumber 243-014-4

RUO – Research Use Only