Learn More
Tingley™ Workbrutes™ Steel-Toe Overshoes
The industry's first injection-molded PVC steel toe overshoe
Supplier: Tingley Rubber Co 35211LG
Description
An affordable way to turn everyday shoes into safety footwear.
- 100% waterproof, seamless construction protects entire shoe from damage caused by site chemicals, contaminants, and wear
- Snug-fitting design and cleated outsole help prevent slips and falls
- Special Frigiflex™ PVC compound provides stretch and longer wear; stays flexible in cold temperatures
- Features include pull-on tab and heel kicker for easy on/off
Specifications
| ASTM F2413-05 M/I/75/C/75. | |
| Shipping, warehousing, baggage handling, food processing, mining, construction, general industry, transportation, onsite visitors, and temporary employees | |
| Large | |
| Chemicals, fats, certain acids, hydrocarbons, caustics |
| Black, Yellow | |
| PVC | |
| Cleated outsole, wedge sole | |
| Yes |