missing translation for 'onlineSavingsMsg'
Learn More

Tingley™ Rubber Work Overboots, 10 in.

Catalog No. 19022888
Change view
Click to view available options
Size:
2X-Large, 12.5 to 14
X-Large, 11 to 12.5
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19022888 X-Large, 11 to 12.5
19022889 2X-Large, 12.5 to 14
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19022888 Supplier Tingley™ Supplier No. 1400 XL

Won't crack or stiffen, even in cold weather

  • Made from exclusive stretch-rubber compound
  • Protect against alcohol, and many organic acids, salts and alkalis
  • Molded rubber button on 10 in. (25.4cm) boot styles for secure closure
  • No hardware to tear out or conduct electricity
  • No lining for easy decontamination
  • Deep, slip-resistant treads
  • Black

Specifications

Closure Type Button
Color Black
For Use With (Application) Food processing, transportation, utilities, construction, general industry, municipalities, and agriculture
Height (English) 10 in.
Material Rubber
Size X-Large, 11 to 12.5
Sole Features Deep-cleated outsole
Resistant To Chemicals, acids, alcohols
Waterproof Yes