Learn More
Tingley™ Rubber Work High-top Galoshes
Won't crack or stiffen, even in cold weather
Supplier: Tingley™ 1300S
Description
- Made from an exclusive stretch-rubber compound
- Protect against alcohol, and many organic acids, salts and alkalis
- No hardware to tear out or conduct electricity
- No lining for easy decontamination
- Deep, slip-resistant treads
- Black
Specifications
| Button | |
| Food processing, transportation, utilities, construction, general industry, municipalities, and agriculture | |
| Small, 6 to 7.5 | |
| High top galoshes | |
| Yes |
| Black | |
| Rubber | |
| Deep-cleated outsole | |
| Chemicals, acids, alcohols |