missing translation for 'onlineSavingsMsg'
Learn More

Tingley™ Rubber Work High-top Galoshes

Catalog No. 19048751
Change view
Click to view available options
Size:
2X-Large, 12.5 to 14
Large, 9.5 to 11
Medium, 8 to 9.5
Small, 6 to 7.5
X-Large, 11 to 12.5
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19048751 Medium, 8 to 9.5
19048753 Large, 9.5 to 11
19051964 X-Large, 11 to 12.5
19051960 2X-Large, 12.5 to 14
19051961 Small, 6 to 7.5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 19048751 Supplier Tingley™ Supplier No. 1300 M

Won't crack or stiffen, even in cold weather

  • Made from an exclusive stretch-rubber compound
  • Protect against alcohol, and many organic acids, salts and alkalis
  • No hardware to tear out or conduct electricity
  • No lining for easy decontamination
  • Deep, slip-resistant treads
  • Black

Specifications

Closure Type Button
Color Black
For Use With (Application) Food processing, transportation, utilities, construction, general industry, municipalities, and agriculture
Material Rubber
Size Medium, 8 to 9.5
Sole Features Deep-cleated outsole
Style High top galoshes
Resistant To Chemicals, acids, alcohols
Waterproof Yes