Learn More
Tingley™ Better Grade PVC Work Boots
Blend of high-quality PVC resins, low-temperature plasticizers, and nitrile rubber creates work boots that remain supple even in cold weather
Supplier:  Tingley™ 005114410
Catalog No. 19022937
Only null left
Explore available promotions
Description
Description
Withstand abrasion, and provide resistance to chemicals such as alcohols, organic and inorganic acids, salts, and alkalis. 100% waterproof. Ideal for agriculture, industrial, and food processing applications.
  • 15 in. high (38cm)
  • Brown upper with crepe sole
  • Cleated outsole spits out debris for excellent gripping action
  • Injection molded construction for 100% waterproof protection
  • Flexible upper material stays supple in cold temperatures to make walking easier
  • Outsole material is soft for excellent slip resistance and absorbs shock when walking
  • Removable polypropylene insole wicks perspiration to help keep feet dry and comfortable
Compliance
Steel-toe work boots meet ANSI Z41 PT99 M/I/75/C/75.
Product Suggestions
Product Suggestions