missing translation for 'onlineSavingsMsg'
Learn More

Tin wire, 1.0mm (0.04in) dia, Puratronic™, 99.998% (metals basis)

Catalog No. AA11905CD
Change view
Click to view available options
Quantity:
50 cm
250 cm
1000 cm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA11905CD 1000 cm
AA11905BW 50 cm
AA11905CB 250 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA11905CD Supplier Thermo Scientific Chemicals Supplier No. 011905CD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Form: approx. 5.72g/m | CAS: 7440-31-5 | Sn | 118.71 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7440-31-5
Molecular Formula Sn
Molecular Weight (g/mol) 118.71
MDL Number MFCD00133862
InChI Key ATJFFYVFTNAWJD-UHFFFAOYSA-N
Synonym powder, stannum, metallic, tin, elemental, wang, zinn, flake, tin, metal, silver matt powder, zinn german
PubChem CID 5352426
ChEBI CHEBI:32990
IUPAC Name tin
SMILES [Sn]

Specifications

Color Silver to Gray
Appearance Silver Gray wire
Quantity 1000 cm
Odor Odorless
Form Wire
Assay Percent Range 99.998% (Metals basis)
Chemical Name or Material Tin
EINECSNumber 23-141-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only