missing translation for 'onlineSavingsMsg'
Learn More

Tin wire, 0.5mm (0.02in) dia, Puratronic™, 99.9985% (metals basis)

Catalog No. AA11906G9
Change view
Click to view available options
Quantity:
1 m
5 m
25 m
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA11906G9 25 m
AA11906G1 1 m
AA11906G5 5 m
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA11906G9 Supplier Thermo Scientific Chemicals Supplier No. 011906G9

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7440-31-5 | Sn | 118.71 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7440-31-5
Molecular Formula Sn
Molecular Weight (g/mol) 118.71
MDL Number MFCD00133862
InChI Key ATJFFYVFTNAWJD-UHFFFAOYSA-N
Synonym powder, stannum, metallic, tin, elemental, wang, zinn, flake, tin, metal, silver matt powder, zinn german
PubChem CID 5352426
ChEBI CHEBI:32990
IUPAC Name tin
SMILES [Sn]

Specifications

Quantity 25 m
Odor Odorless
Weight ≈1.43g/m
Form Wire
Percent Purity 99.9985%
Grade Puratronic
Assay (metals basis)
Chemical Name or Material Tin wire, 0.5mm (0.02 in.) dia.
EINECSNumber 23-141-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only