missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tin wire, 0.25mm (0.01in) dia, Puratronic™, 99.998% (metals basis)
Form: Approx. 0.36g/m | CAS: 7440-31-5 | Sn | 118.71 g/mol
Supplier: Thermo Scientific Chemicals 011907G5
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Silver to Gray | |
| 7440-31-5 | |
| 7.31g/cm3 | |
| Odorless | |
| Wire | |
| MFCD00133862 | |
| ATJFFYVFTNAWJD-UHFFFAOYSA-N | |
| tin | |
| 5352426 |
| Tin | |
| Silver Gray wire | |
| 5 m | |
| 99.998% (Metals basis) | |
| Sn | |
| powder, stannum, metallic, tin, elemental, wang, zinn, flake, tin, metal, silver matt powder, zinn german | |
| [Sn] | |
| 118.71 | |
| CHEBI:32990 |
Chemical Identifiers
| 7440-31-5 | |
| 118.71 | |
| ATJFFYVFTNAWJD-UHFFFAOYSA-N | |
| 5352426 | |
| tin |
| Sn | |
| MFCD00133862 | |
| powder, stannum, metallic, tin, elemental, wang, zinn, flake, tin, metal, silver matt powder, zinn german | |
| CHEBI:32990 | |
| [Sn] |
Safety and Handling
EINECSNumber : 23-141-8
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only