missing translation for 'onlineSavingsMsg'
Learn More

Tin slug, 11mm (0.43 in.) dia. x 2mm (0.1 in.) thick, Puratronic™, 99.995% (metals basis), Thermo Scientific™

Catalog No. AA41169KD
Change view
Click to view available options
Quantity:
50 Pc.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA41169KD 50 Pc.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA41169KD Supplier Thermo Scientific Chemicals Supplier No. 41169KD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Evaporation source

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 7440-31-5
Molecular Formula Sn
Molecular Weight (g/mol) 118.71
MDL Number MFCD00133862
InChI Key ATJFFYVFTNAWJD-UHFFFAOYSA-N
Synonym powder, stannum, metallic, tin, elemental, wang, zinn, flake, tin, metal, silver matt powder, zinn german
PubChem CID 5352426
ChEBI CHEBI:32990
IUPAC Name tin
SMILES [Sn]

Specifications

Quantity 50 Pc.
Odor Odorless
Weight ≈1.3g
Form Slug
Percent Purity 99.995%
Grade Puratronic
Assay (metals basis)
Chemical Name or Material Tin slug, 11mm (0.43 in.) dia. x 2mm (0.1 in.) thick
EINECSNumber 231-141-8
TSCA Yes
Recommended Storage Ambient temperatures