missing translation for 'onlineSavingsMsg'
Learn More

Tin shot, 8-20 mesh, 99.8% (metals basis)

Catalog No. p-4407987
Change view
Click to view available options
Quantity:
100 g
500 g
2 kg
Catalog No. Quantity
AA1037722 100 g
AA1037736 500 g
AA10377A3 2 kg
3 options

Catalog No. AA1037722

Supplier: Thermo Scientific Chemicals 01037722

Only null left

CAS: 7440-31-5 | Sn | 118.71 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7440-31-5
Molecular Formula Sn
Molecular Weight (g/mol) 118.71
MDL Number MFCD00133862
InChI Key ATJFFYVFTNAWJD-UHFFFAOYSA-N
Synonym powder, stannum, metallic, tin, elemental, wang, zinn, flake, tin, metal, silver matt powder, zinn german
PubChem CID 5352426
ChEBI CHEBI:32990
IUPAC Name tin
SMILES [Sn]

Specifications

Color Silver to Gray
Appearance Silver Gray shot
Quantity 100 g
Mesh Size 8-20. Mesh
Odor Odorless
Form Shot
Assay Percent Range 99.8% (Metals basis)
Chemical Name or Material Tin
EINECSNumber 231-141-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only