missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tin shot, 3mm (0.1in), 99.999% (metals basis)
CAS: 7440-31-5 | Sn | 118.71 g/mol
$236.55 - $2598.53
Chemical Identifiers
| CAS | 7440-31-5 |
|---|---|
| Molecular Formula | Sn |
| Molecular Weight (g/mol) | 118.71 |
| MDL Number | MFCD00133862 |
| InChI Key | ATJFFYVFTNAWJD-UHFFFAOYSA-N |
| Synonym | powder, stannum, metallic, tin, elemental, wang, zinn, flake, tin, metal, silver matt powder, zinn german |
| PubChem CID | 5352426 |
| ChEBI | CHEBI:32990 |
| IUPAC Name | tin |
| SMILES | [Sn] |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AA1101014
|
Thermo Scientific Chemicals
01101014 |
25 g |
Each for $236.55
|
|
|||||
|
AA1101022
|
Thermo Scientific Chemicals
01101022 |
100 g |
Each for $679.64
|
|
|||||
|
AA1101036
|
Thermo Scientific Chemicals
01101036 |
500 g |
Each for $2,598.53
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 7440-31-5 | |
| 118.71 | |
| ATJFFYVFTNAWJD-UHFFFAOYSA-N | |
| 5352426 | |
| tin |
| Sn | |
| MFCD00133862 | |
| powder, stannum, metallic, tin, elemental, wang, zinn, flake, tin, metal, silver matt powder, zinn german | |
| CHEBI:32990 | |
| [Sn] |
Specifications
| Silver to Gray | |
| Silver Gray shot | |
| Odorless | |
| Shot | |
| MFCD00133862 | |
| ATJFFYVFTNAWJD-UHFFFAOYSA-N | |
| tin | |
| 5352426 | |
| Tin |
| 7440-31-5 | |
| 25 g | |
| 99.999% (Metals basis) | |
| Sn | |
| powder, stannum, metallic, tin, elemental, wang, zinn, flake, tin, metal, silver matt powder, zinn german | |
| [Sn] | |
| 118.71 | |
| CHEBI:32990 |
Safety and Handling
EINECSNumber : 231-141-8
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only