missing translation for 'onlineSavingsMsg'
Learn More

Tin rod, 13mm (0.51in) dia, Puratronic™, 99.998+% (metals basis)

Catalog No. AA0078806
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA0078806 5 g
AA0078814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA0078806 Supplier Thermo Scientific Chemicals Supplier No. 00078806

CAS: 7440-31-5 | Sn | 118.71 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7440-31-5
Molecular Formula Sn
Molecular Weight (g/mol) 118.71
MDL Number MFCD00133862
InChI Key ATJFFYVFTNAWJD-UHFFFAOYSA-N
Synonym powder, stannum, metallic, tin, elemental, wang, zinn, flake, tin, metal, silver matt powder, zinn german
PubChem CID 5352426
ChEBI CHEBI:32990
IUPAC Name tin
SMILES [Sn]

Specifications

Quantity 5 g
Odor Odorless
Weight ≈0.52cm/5g
Form Rod
Percent Purity ≥99.998%
Grade Puratronic
Assay (metals basis)
Chemical Name or Material Tin rod, 13mm (0.51 in.) dia.
EINECSNumber 231-141-8
RTECSNumber XP7320000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only