missing translation for 'onlineSavingsMsg'
Learn More

Tin powder, spherical, APS approximately 0.1 micron

Catalog No. AA4346122
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4346122 100 g
AA4346114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4346122 Supplier Thermo Scientific Chemicals Supplier No. 04346122

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7440-31-5 | Sn | 118.71 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7440-31-5
Molecular Formula Sn
Molecular Weight (g/mol) 118.71
MDL Number MFCD00133862
InChI Key ATJFFYVFTNAWJD-UHFFFAOYSA-N
Synonym powder, stannum, metallic, tin, elemental, wang, zinn, flake, tin, metal, silver matt powder, zinn german
PubChem CID 5352426
ChEBI CHEBI:32990
IUPAC Name tin
SMILES [Sn]

Specifications

Quantity 100 g
Odor Odorless
Particle Size APS ≈ 0.1 Micron
Form Spherical Powder
UN Number UN3089
Chemical Name or Material Tin powder
Hazard Category H302-H319-H335-H336-H373-H500
Hazard Statement GHS H Statement
H228
Flammable solid.
Precautionary Statement P260-P264b-P270-P271-P280i-P301+P312-P304+P340-P305+P351+P338-P312-P314-P330-P501c
DOTInformation Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: METAL POWDERS, FLAMMABLE, N.O.S.
EINECSNumber 231-141-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only