Learn More
Tin powder, -100 mesh, 99.995% (metals basis)
CAS: 7440-31-5 | Sn | 118.71 g/mol
Supplier: Thermo Scientific Chemicals 01101309
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Gray | |
| 7440-31-5 | |
| 7.31g/cm3 | |
| -100 Mesh | |
| Powder | |
| Sn | |
| powder, stannum, metallic, tin, elemental, wang, zinn, flake, tin, metal, silver matt powder, zinn german | |
| [Sn] | |
| 118.71 | |
| CHEBI:32990 |
| Tin | |
| Gray powder | |
| 10 g | |
| Odorless | |
| 99.995% | |
| MFCD00133862 | |
| ATJFFYVFTNAWJD-UHFFFAOYSA-N | |
| tin | |
| 5352426 |
Chemical Identifiers
| 7440-31-5 | |
| 118.71 | |
| ATJFFYVFTNAWJD-UHFFFAOYSA-N | |
| 5352426 | |
| tin |
| Sn | |
| MFCD00133862 | |
| powder, stannum, metallic, tin, elemental, wang, zinn, flake, tin, metal, silver matt powder, zinn german | |
| CHEBI:32990 | |
| [Sn] |
Safety and Handling
P260-P264b-P270-P271-P280i-P301+P312-P304+P340-P305+P351+P338-P312-P314-P330-P501c
H302-H319-H335-H336-H373-H500
EINECSNumber : 231-141-8
RTECSNumber : XP7320000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only