missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tin(IV) phthalocyanine dichloride
Description
Tin(IV) phthalocyanine dichloride is a stable metal phthalocyanine dye utilized in biochemical research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Quantity | 1 g |
| UN Number | UN3146 |
| Sensitivity | Moisture sensitive |
| Solubility Information | Insoluble in water. |
| Formula Weight | 702.34 |
| Physical Form | Powder |
| Chemical Name or Material | Tin(IV) phthalocyanine dichloride |
Safety and Handling
| Hazard Category | H301 |
| Hazard Statement | GHS H Statement H301 Toxic if swallowed. |
| Precautionary Statement | P264b-P270-P301+P310-P330-P501c |
| DOTInformation | Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOTIN COMPOUNDS, SOLID, N.O.S. |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only