missing translation for 'onlineSavingsMsg'
Learn More

Tin(IV) phthalocyanine dichloride

Catalog No. AA3960606
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3960606 5 g
AA3960603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3960606 Supplier Thermo Scientific Chemicals Supplier No. 03960606

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 18253-54-8 | C32H16Cl2N8Sn | 702.146 g/mol

Tin(IV) phthalocyanine dichloride is a stable metal phthalocyanine dye utilized in biochemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 18253-54-8
Molecular Formula C32H16Cl2N8Sn
Molecular Weight (g/mol) 702.146
MDL Number MFCD00054210
InChI Key BBRNKSXHHJRNHK-UHFFFAOYSA-L
Synonym tin iv phthalocyanine dichloride, sncl2pc, dichlorotin phthalocyanine, tin phthalocyanine dichloride, tin iv phthalocyaninedichloride
PubChem CID 9580361
SMILES C1=CC=C2C(=C1)C3=NC4=C5C=CC=CC5=C6N4[Sn](N7C(=NC2=N3)C8=CC=CC=C8C7=NC9=NC(=N6)C1=CC=CC=C19)(Cl)Cl

Specifications

Quantity 5 g
UN Number UN3146
Sensitivity Moisture sensitive
Solubility Information Insoluble in water.
Formula Weight 702.34
Physical Form Powder
Chemical Name or Material Tin(IV) phthalocyanine dichloride
Hazard Category H301
Hazard Statement GHS H Statement
H301
Toxic if swallowed.
Precautionary Statement P264b-P270-P301+P310-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOTIN COMPOUNDS, SOLID, N.O.S.
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only