missing translation for 'onlineSavingsMsg'
Learn More

Tin(IV) Phthalocyanine Dichloride, TCI America™

Catalog No. P09971G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P09971G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P09971G Supplier TCI America Supplier No. P09971G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 18253-54-8
Molecular Formula C32H16Cl2N8Sn
Molecular Weight (g/mol) 702.146
MDL Number MFCD00054210
InChI Key BBRNKSXHHJRNHK-UHFFFAOYSA-L
Synonym tin iv phthalocyanine dichloride, sncl2pc, dichlorotin phthalocyanine, tin phthalocyanine dichloride, tin iv phthalocyaninedichloride
PubChem CID 9580361
SMILES C1=CC=C2C(=C1)C3=NC4=C5C=CC=CC5=C6N4[Sn](N7C(=NC2=N3)C8=CC=CC=C8C7=NC9=NC(=N6)C1=CC=CC=C19)(Cl)Cl

Specifications

Color Blue-Purple
Physical Form Crystalline Powder
Quantity 1 g
UN Number 3146
Formula Weight 702.15
Chemical Name or Material Tin(IV) Phthalocyanine Dichloride