missing translation for 'onlineSavingsMsg'
Learn More

Tin(IV) oxide, Puratronic™, 99.996% (metals basis)

Catalog No. AA1089106
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1089106 5 g
AA1089114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1089106 Supplier Thermo Scientific Chemicals Supplier No. 01089106

CAS: 18282-10-5 | O2Sn | 150.71 g/mol

Tin(IV) oxide is used as a pigment in the ceramic industry and as a polishing powder. It finds use in sensors of combustible gases. It is also used as a catalyst along with vanadium oxide for the oxidation of aromatic compounds to prepare carboxylic acids and acid anhydrides. It is used in sensors for detection of trace levels of carbon monoxide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 18282-10-5
Molecular Formula O2Sn
Molecular Weight (g/mol) 150.71
MDL Number MFCD00011244
InChI Key XOLBLPGZBRYERU-UHFFFAOYSA-N
Synonym tin dioxide, tin iv oxide, stannic oxide, cassiterite, flowers of tin, stannic dioxide, tin oxide sno2, tin peroxide, white tin oxide, stannic anhydride
PubChem CID 29011
ChEBI CHEBI:52991
IUPAC Name dioxotin
SMILES O=[Sn]=O

Specifications

Boiling Point 1800°C to 1900°C (sublimation)
Melting Point 1630°C
Physical Form Powder
Quantity 5 g
Assay Percent Range 99.996% (metals basis)
Solubility Information Soluble in strong acids and alkali. Insoluble in water.Soluble in hot concentrated alkali and concentrated acids. Insoluble in water and alcohols.
Formula Weight 150.69
Refractive Index 1.9968
Density 6.95 g/mL
Chemical Name or Material Tin(IV) oxide
EINECSNumber 242-159-0
RTECSNumber XQ4000000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only