missing translation for 'onlineSavingsMsg'
Learn More

Tin(IV) Oxide, Extra Pure, MilliporeSigma™

Catalog No. p-4939068
Change view
Click to view available options
Quantity:
250 g
25 kg
Packaging:
Fibre carton
Plastic bottle
Catalog No. Quantity Packaging
M1078180250 250 g Plastic bottle
M1078189025 25 kg Fibre carton
2 options

Catalog No. M1078180250

Supplier: Technidata 1.07818.0250

Only null left

Chemical Identifiers

CAS 18282-10-5
Molecular Formula O2Sn
Molecular Weight (g/mol) 150.71
MDL Number MFCD00011244
InChI Key XOLBLPGZBRYERU-UHFFFAOYSA-N
Synonym tin dioxide, tin iv oxide, stannic oxide, cassiterite, flowers of tin, stannic dioxide, tin oxide sno2, tin peroxide, white tin oxide, stannic anhydride
PubChem CID 29011
ChEBI CHEBI:52991
IUPAC Name stannanedione
SMILES O=[Sn]=O

Specifications

Physical Form Solid
Quantity 250 g
Formula Weight 150.69
Grade Extra Pure
Odor Odorless
Packaging Plastic bottle
Appearance Off-white solid
Chemical Name or Material Tin(IV) oxide