missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tin(IV) Oxide, Extra Pure, MilliporeSigma™
Specifications
Specifications
| Physical Form | Solid |
| Quantity | 25 kg |
| Odor | Odorless |
| Packaging | Fibre carton |
| Appearance | Off-white solid |
| Formula Weight | 150.69 |
| Percent Purity | ≥99.0% |
| Grade | Extra Pure |
| Chemical Name or Material | Tin(IV) oxide |