missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tin(IV) oxide, 99%, pure
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Quantity | 100 g |
| Specific Gravity | 6.95 |
| Density | 6.9500g/mL |
| Linear Formula | SnO2 |
| Merck Index | 15,8905 |
| Solubility Information | Solubility in water: insoluble. Other solubilities: slowly soluble in sodium hydroxide solution, insoluble in alcohol and cold acids |
| Formula Weight | 150.69 |
| Percent Purity | 98.5% min. (metals basis) |
| Grade | Pure |
| Chemical Name or Material | Tin(IV) oxide |
Safety and Handling
| EINECSNumber | 242-159- |
RUO – Research Use Only