missing translation for 'onlineSavingsMsg'
Learn More

Tin(IV) oxide, 99%, pure

Catalog No. AC451810025
Change view
Click to view available options
Quantity:
100 g
1 kg
2.5 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC451810025 2.5 kg
AC451811000 100 g
AC451810010 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC451810025 Supplier Thermo Scientific Chemicals Supplier No. 451810025

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 18282-10-5 | O2Sn | 150.71 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 18282-10-5
Molecular Formula O2Sn
Molecular Weight (g/mol) 150.71
MDL Number MFCD00011244
InChI Key XOLBLPGZBRYERU-UHFFFAOYSA-N
Synonym tin dioxide, tin iv oxide, stannic oxide, cassiterite, flowers of tin, stannic dioxide, tin oxide sno2, tin peroxide, white tin oxide, stannic anhydride
PubChem CID 29011
ChEBI CHEBI:52991
IUPAC Name dioxotin
SMILES O=[Sn]=O

Specifications

Quantity 2.5 kg
Specific Gravity 6.95
Density 6.9500g/mL
Linear Formula SnO2
Merck Index 15,8905
Solubility Information Solubility in water: insoluble. Other solubilities: slowly soluble in sodium hydroxide solution, insoluble in alcohol and cold acids
Formula Weight 150.69
Percent Purity 99%
Grade Pure
Chemical Name or Material Tin(IV) oxide
EINECSNumber 242-159-

RUO – Research Use Only