missing translation for 'onlineSavingsMsg'
Learn More

Tin(IV) oxide, 99.9%, (trace metal basis)

Catalog No. AC223681000
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC223681000 100 g Plastic bottle
AC223685000 500 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC223681000 Supplier Thermo Scientific Chemicals Supplier No. 223681000

CAS: 18282-10-5 | O2Sn | 150.71 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For ultra trace analysis

Chemical Identifiers

CAS 18282-10-5
Molecular Formula O2Sn
Molecular Weight (g/mol) 150.71
MDL Number MFCD00011244
InChI Key XOLBLPGZBRYERU-UHFFFAOYSA-N
Synonym tin dioxide, tin iv oxide, stannic oxide, cassiterite, flowers of tin, stannic dioxide, tin oxide sno2, tin peroxide, white tin oxide, stannic anhydride
PubChem CID 29011
ChEBI CHEBI:52991
IUPAC Name stannanedione
SMILES O=[Sn]=O

Specifications

Color White to Gray
Physical Form Powder
Quantity 100 g
Assay Percent Range 99.9%
Linear Formula SnO2
Merck Index 15, 8905
Solubility Information Solubility in water: insoluble. Other solubilities: slowly soluble in sodium hydroxide solution, insoluble in alcohol and cold acids
Formula Weight 150.69
Percent Purity 99.9%
Grade Trace Metal Basis
Packaging Plastic bottle
Specific Gravity 6.95
Density 6.9500g/mL
Chemical Name or Material Tin(IV) oxide, 99.9%
Show More Show Less
EINECSNumber 242-159-0
RTECSNumber XQ4000000
TSCA TSCA

RUO – Research Use Only