Learn More
Tin(IV) oxide, 99.9% (metals basis)
Description
For vacuum deposition. Used as a catalyst, in putty, as a polishing powder for steel and glass, in ceramic glazes and colors. The electrical properties of tin (iv) oxide render it an excellent material for the detection of very low levels of carbon monoxide, and several devices based on sintered pellets of SnO 2 are commercially available. Dye-sensitized fully solid state cells having the structure of rough n-type semiconductor film comprising zinc and tin (IV) oxide/Ru-bipyridyl complex/p-CuI are found to generate high short-circuit photocurrents and open-circuit voltages. The preparation of hydrous tin (IV) oxide in a suitable form for use as an ion exchanger is described. a colloidal-crystal templating technique to prepare three-dimensionally ordered macroporous (3DOM) tin (iv) oxide films for use as lithium ion anodes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Boiling Point | 1800°C to 1900°C (sublimation) |
| Melting Point | 1630°C |
| Color | White |
| Physical Form | Sintered Lump |
| Quantity | 100 g |
| Odor | Odorless |
| Refractive Index | 1.9968 |
| Density | 6.95 g/mL |
| Linear Formula | SnO2 |
| Merck Index | 14,8778 |
| Show More |
Safety and Handling
| EINECSNumber | 242-159-0 |
| RTECSNumber | XQ4000000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only