missing translation for 'onlineSavingsMsg'
Learn More

Tin(IV) isopropoxide isopropanol adduct, 98% (metals basis)

Catalog No. AA4126906
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4126906 5 g
AA4126903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4126906 Supplier Thermo Scientific Chemicals Supplier No. 04126906

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 132951-93-0

Tin(IV) isopropoxide isopropanol adduct is used has a important tin precursor for the synthesis of tin oxide nanoparticles. Used in Indium Tin Oxide films, electrochemical and conducting properties and thin films.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 132951-93-0
MDL Number MFCD00145408

Specifications

Melting Point ∼130°C (decomposition)
Boiling Point 131°C (1.6 mmHg)
Quantity 5 g
Sensitivity Moisture sensitive
Solubility Information Soluble in hydrocarbons and warm isopropanol. Hydrolyzes with water. Reacts with alcohols,ketones and esters.
Formula Weight 414.15
Percent Purity 98%
Physical Form Wet Solid
Assay (metals basis)
Chemical Name or Material Tin(IV) isopropoxide isopropanol adduct
Hazard Category H315-H318-H335
Hazard Statement GHS H Statement
H318-H335-H315
Causes serious eye damage.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P310-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only