Learn More
Tin(IV) isopropoxide, 99% (metals basis), 10% w/v in isopropanol
CAS: 1184-61-8 | C12H32O4Sn | 359.094 g/mol
$73.42 - $2759.18
Chemical Identifiers
| CAS | 1184-61-8 |
|---|---|
| Molecular Formula | C12H32O4Sn |
| Molecular Weight (g/mol) | 359.094 |
| MDL Number | MFCD00145408 |
| InChI Key | VUDAJANAMZGVOP-UHFFFAOYSA-N |
| Synonym | tin iv isopropoxide, tin tetraisopropoxide, acmc-20aji2, tin 4+ tetrapropan-2-olate, tin 4+ ion tetrakis propan-2-olate, tin 4+ tetrakis propan-2-olate |
| PubChem CID | 85354423 |
| IUPAC Name | propan-2-ol;tin |
| SMILES | CC(C)O.CC(C)O.CC(C)O.CC(C)O.[Sn] |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AA45480AA
|
Thermo Scientific Chemicals
045480AA |
5 mL |
Each for $73.42
|
|
|||||
|
AA45480AC
|
Thermo Scientific Chemicals
045480AC |
25 mL |
Each for $247.14
|
|
|||||
|
AA45480AE
|
Thermo Scientific Chemicals
045480AE |
100 mL |
Each for $656.70
|
|
|||||
|
AA45480AP
|
Thermo Scientific Chemicals
045480AP |
500 mL |
Each for $2,759.18
|
|
|||||
Description
Tin(IV) isopropoxide is used as a precursor for the preparation of tin oxide nanoparticles, thin films, indium tin oxide (ITO) films , which shows desired electrochemical and conducting properties. Doped tin oxide nanoparticles and thin films find application as gas sensors.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 1184-61-8 | |
| 359.094 | |
| VUDAJANAMZGVOP-UHFFFAOYSA-N | |
| 85354423 | |
| CC(C)O.CC(C)O.CC(C)O.CC(C)O.[Sn] |
| C12H32O4Sn | |
| MFCD00145408 | |
| tin iv isopropoxide, tin tetraisopropoxide, acmc-20aji2, tin 4+ tetrapropan-2-olate, tin 4+ ion tetrakis propan-2-olate, tin 4+ tetrakis propan-2-olate | |
| propan-2-ol;tin |
Specifications
| 1184-61-8 | |
| Alcohol-like | |
| Sn[OCH(CH3)2]4 | |
| 5 mL | |
| Moisture sensitive | |
| Miscible with alcohols,ketones and esters. | |
| CC(C)O.CC(C)O.CC(C)O.CC(C)O.[Sn] | |
| 359.094 | |
| 355.06 | |
| Liquid | |
| (metals basis) |
| Colorless | |
| C12H32O4Sn | |
| MFCD00145408 | |
| UN3274 | |
| tin iv isopropoxide, tin tetraisopropoxide, acmc-20aji2, tin 4+ tetrapropan-2-olate, tin 4+ ion tetrakis propan-2-olate, tin 4+ tetrakis propan-2-olate | |
| VUDAJANAMZGVOP-UHFFFAOYSA-N | |
| propan-2-ol;tin | |
| 85354423 | |
| 99% | |
| Tin (IV) isopropoxide |
Safety and Handling
GHS H Statement
H225-H314-H302-H336
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Harmful if swallowed.
May cause drowsiness or dizziness.
P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H225-H314-H335-H336
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ALCOHOLATES SOLUTION, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only