missing translation for 'onlineSavingsMsg'
Learn More

Tin(IV) isopropoxide, 98% (metals basis), 10% w/v in isopropanol/toluene

Catalog No. AA36563AE
Change view
Click to view available options
Quantity:
5 mL
25 mL
100 mL
500 mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA36563AE 100 mL
AA36563AA 5 mL
AA36563AC 25 mL
AA36563AP 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AA36563AE Supplier Thermo Scientific Chemicals Supplier No. 036563AE
Only null left

CAS: 1184-61-8 | C12H32O4Sn | 359.094 g/mol

Tin(IV) isopropoxide isopropanol is used has a important tin precursor for the synthesis of tin oxide nanoparticles. Used in Indium Tin Oxide films, thin films doped tin oxide nanoparticles, thin films and are widely studied as gas sensors. Tin(IV) precursor solution was prepared from tin(IV) isopropoxide dissolved in isopropyl alcohol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1184-61-8
Molecular Formula C12H32O4Sn
Molecular Weight (g/mol) 359.094
MDL Number MFCD00145408
InChI Key VUDAJANAMZGVOP-UHFFFAOYSA-N
Synonym tin iv isopropoxide, tin tetraisopropoxide, acmc-20aji2, tin 4+ tetrapropan-2-olate, tin 4+ ion tetrakis propan-2-olate, tin 4+ tetrakis propan-2-olate
PubChem CID 85354423
IUPAC Name propan-2-ol;tin
SMILES CC(C)O.CC(C)O.CC(C)O.CC(C)O.[Sn]

Specifications

Quantity 100 mL
UN Number UN1993
Sensitivity Moisture sensitive
Solubility Information Soluble in hydrocarbons,reacts with alcohols,ketones and esters
Formula Weight 355.06
Percent Purity 98%
Physical Form Liquid
Assay (metals basis)
Chemical Name or Material Tin(IV) isopropoxide
Hazard Category H225-H304-H314-H335-H336-H361d-H373
Hazard Statement GHS H Statement
H225-H304-H361-H373-H314-H336
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.
Suspected of damaging fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Causes severe skin burns and eye damage.
May cause drowsiness or dizziness.
Precautionary Statement P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P281-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only