missing translation for 'onlineSavingsMsg'
Learn More

Tin(IV) iodide, 99+%, -10 mesh, Thermo Scientific™

Chemical Identifiers

CAS 7790-47-8
Molecular Formula H4I4Sn
Molecular Weight (g/mol) 630.36
MDL Number MFCD00036272
InChI Key ZBCVLXKKLMNLGV-UHFFFAOYSA-N
Synonym stannic iodide, stannane, tetraiodo, tin iv iodide, tetraiodotin, tin tetraiodide, tin iodide, unii-8h688bw69x, tin iv iodide 1:4, tin-iv-iodide
PubChem CID 24631
IUPAC Name tetraiodostannane
SMILES [Sn].I.I.I.I
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC388560100
SDS View Documents
Thermo Scientific Chemicals
388560100
10g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC388560500
SDS View Documents
Thermo Scientific Chemicals
388560500
50g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Lewis acid
Specifications

Chemical Identifiers

7790-47-8
630.36
ZBCVLXKKLMNLGV-UHFFFAOYSA-N
24631
[Sn].I.I.I.I
H4I4Sn
MFCD00036272
stannic iodide, stannane, tetraiodo, tin iv iodide, tetraiodotin, tin tetraiodide, tin iodide, unii-8h688bw69x, tin iv iodide 1:4, tin-iv-iodide
tetraiodostannane

Specifications

7790-47-8
99% min. (Argentometry)
SnI4
15, 8904
Solubility in water: decomposes. Other solubilities: soluble in alcohol,benzene,chloroform,ether and,carbon disulfide
[Sn].I.I.I.I
630.36
626.31
Glass bottle
10g
H4I4Sn
MFCD00036272
stannic iodide, stannane, tetraiodo, tin iv iodide, tetraiodotin, tin tetraiodide, tin iodide, unii-8h688bw69x, tin iv iodide 1:4, tin-iv-iodide
ZBCVLXKKLMNLGV-UHFFFAOYSA-N
tetraiodostannane
24631
99+%
Tin(IV) iodide
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 232-208-4

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only