missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tin(IV) chloride, 1M solution in dichloromethane, AcroSeal™
CAS: 7646-78-8 | Cl4Sn | 260.52 g/mol
$122.07 - $122.07
Specifications
| Chemical Name or Material | Tin(IV) chloride |
|---|---|
| Name Note | 1M Solution in Dichloromethane |
| CAS | 7646-78-8,75-09-2 |
| Molecular Formula | Cl4Sn |
| MDL Number | MFCD00011242 |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC378201000
|
Thermo Scientific Chemicals
378201000 |
100 mL | AcroSeal™ Glass bottle |
Each for $122.07
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Lewis acidSpecifications
| Tin(IV) chloride | |
| 7646-78-8,75-09-2 | |
| MFCD00011242 | |
| 15, 8901 | |
| Solubility in water: reacts | |
| Cl[Sn](Cl)(Cl)Cl | |
| 260.52 | |
| 260.52 | |
| Clear colorless solution, fuming in air | |
| 1.4190g/mL |
| 1M Solution in Dichloromethane | |
| Cl4Sn | |
| 09,436; 17,335; 14,304; 13,300; 12,486; 10,370; 07,342; 06,553; 05,627; 03,269; 01,1111; 11,552 | |
| tin tetrachloride, tin iv chloride, stannane, tetrachloro, tin chloride, stannic chloride, tetrachlorotin, tin perchloride, tintetrachloride, tin iv tetrachloride, sncl4 | |
| HPGGPRDJHPYFRM-UHFFFAOYSA-J | |
| tetrachlorostannane | |
| 24287 | |
| 1M, exact strength on the certificate of analysis | |
| 1.419 |
RUO – Research Use Only