missing translation for 'onlineSavingsMsg'
Learn More

Tin(IV) bromide, 99%

Catalog No. p-7080309
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA7112018 50 g
AA7112030 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA7112018 Supplier Thermo Scientific Chemicals Supplier No. 07112018

CAS: 7789-67-5 | Br4Sn | 438.326 g/mol

A specialty product for proteomics research. Used as a precursor for SnO2 through laser-assisted CVD with higher efficiency than other precursors such as dibutyl tin diacetate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7789-67-5
Molecular Formula Br4Sn
Molecular Weight (g/mol) 438.326
MDL Number MFCD00011240
InChI Key LTSUHJWLSNQKIP-UHFFFAOYSA-J
Synonym tin iv bromide, stannic bromide, tin tetrabromide, stannane, tetrabromo, tin perbromide, snbr4, tin bromide, tin iv bromide 1:4, tetrabromotin
PubChem CID 24616
IUPAC Name tetrabromostannane
SMILES Br[Sn](Br)(Br)Br

Specifications

Boiling Point 202°C (734mmHg)
Melting Point 31°C
Color Colorless to White
Physical Form Lump
Quantity 50 g
UN Number UN3260
Merck Index 14,8773
Solubility Information Very soluble in water. Soluble in ethyl alcohol.
Formula Weight 438.33
Percent Purity 99%
Odor Acidic
Sensitivity Air and moisture sensitive
Density 3.34 g/mL
Chemical Name or Material Tin (IV) bromide
Show More Show Less
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318-H335
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause respiratory irritation.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.
EINECSNumber 232-184-5
RTECSNumber XP8300000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only