missing translation for 'onlineSavingsMsg'
Learn More

Tin(IV) 2-ethylhexanoate diisopropoxide, 98% (metals basis)

Catalog No. AA3916906
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3916906 5 g
AA3916903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3916906 Supplier Thermo Scientific Chemicals Supplier No. 03916906

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 288374-32-3

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 288374-32-3
MDL Number MFCD00210604
Synonym tin iv 2-ethylhexanoate diisopropoxide, tin iv 2-ethylhexanoatediisopropoxide metalsbasis

Specifications

Quantity 5 g
Sensitivity Moisture sensitive
Formula Weight 523.08
Percent Purity 98%
Physical Form Liquid
Assay (metals basis)
Chemical Name or Material Tin(IV) 2-ethylhexanoate diisopropoxide
Hazard Category H315-H319-H361d
Hazard Statement GHS H Statement
H361-H315-H319
Suspected of damaging fertility or the unborn child.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P201-P202-P264b-P280i-P281-P302+P352-P305+P351+P338-P308+P313-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only