missing translation for 'onlineSavingsMsg'
Learn More

Tin(II) Trifluoromethanesulfonate 90.0+%, TCI America™

Catalog No. T11941G
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T11941G 1 g
T11945G 5 g
T119425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T11941G Supplier TCI America Supplier No. T11941G

Chemical Identifiers

CAS 62086-04-8
Molecular Formula CH4F3O3SSn+
Molecular Weight (g/mol) 271.805
MDL Number MFCD00191251
InChI Key IACSYIHNPIQPMP-UHFFFAOYSA-M
Synonym tin ii trifluoromethanesulfonate
PubChem CID 131676109
IUPAC Name molecular hydrogen;tin(2+);trifluoromethanesulfonate
SMILES [HH].[HH].C(F)(F)(F)S(=O)(=O)[O-].[Sn+2]

Specifications

Color White-Yellow
Physical Form Crystalline Powder
Quantity 1 g
UN Number 3146
Formula Weight 416.84
Percent Purity ≥90.0% (T)
Chemical Name or Material Tin(II) Trifluoromethanesulfonate