missing translation for 'onlineSavingsMsg'
Learn More

Tin(II) trifluoromethanesulfonate, 98%

Catalog No. AC312530010
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC312530010 1 g Plastic bottle
AC312530050 5 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC312530010 Supplier Thermo Scientific Chemicals Supplier No. 312530010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 62086-04-8 | C2F6O6S2Sn | 416.85 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 62086-04-8
Molecular Formula C2F6O6S2Sn
Molecular Weight (g/mol) 416.85
MDL Number MFCD00191251
InChI Key IACSYIHNPIQPMP-UHFFFAOYSA-M
Synonym tin ii trifluoromethanesulfonate
PubChem CID 131676109
IUPAC Name molecular hydrogen;tin(2+);trifluoromethanesulfonate
SMILES [HH].[HH].C(F)(F)(F)S(=O)(=O)[O-].[Sn+2]

Specifications

Melting Point >300.0°C
Color White to Yellow
Physical Form Powder
Quantity 1 g
Assay Percent Range 97.5% min. (ex Sn)
Solubility Information Solubility in water: soluble
Formula Weight 416.85
Percent Purity 98%
Infrared Spectrum Authentic
Packaging Plastic bottle
Chemical Name or Material Tin(II) trifluoromethanesulfonate
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi

RUO – Research Use Only