missing translation for 'onlineSavingsMsg'
Learn More

Tin(II) sulfate, 95%

Catalog No. AC223665000
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC223665000 500 g Glass bottle
AC223661000 100 g Glass bottle
AC223660025 2.5 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC223665000 Supplier Thermo Scientific Chemicals Supplier No. 223665000

CAS: 7488-55-3 | O4SSn | 214.77 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7488-55-3
Molecular Formula O4SSn
Molecular Weight (g/mol) 214.77
MDL Number MFCD00011246
InChI Key OBBXFSIWZVFYJR-UHFFFAOYSA-L
Synonym stannous sulfate, tin ii sulfate, tin 2+ sulfate, unii-0mfe10j96e, sulfuric acid, tin 2+ salt 1:1, stannous sulfate, crystal, stannous sulfate, solutions, tin, ion sn2+ sulfate, tin ii sulfate 250g
PubChem CID 62643
SMILES [Sn++].[O-]S([O-])(=O)=O

Specifications

Melting Point 360.0°C
Color White
Physical Form Powder, Crystals and/or Lumps
Quantity 500 g
Assay Percent Range 94% min. (Iodimetry)
Linear Formula SnSO4
Merck Index 15,8917
Solubility Information Solubility in water: soluble. Other solubilities: soluble in diluted sulfuric acid
IUPAC Name tin(2+);sulfate
Formula Weight 214.75
Percent Purity 95%
Packaging Glass bottle
Chemical Name or Material Tin(II) sulfate
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes skin irritation.
May cause an allergic skin reaction.
Causes serious eye damage.
Harmful if inhaled.
May cause respiratory irritation.
May cause damage to organs through prolonged or repeated ex
Precautionary Statement GHS P Statement
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minu
EINECSNumber 231-302-2

RUO – Research Use Only