missing translation for 'onlineSavingsMsg'
Learn More

Tin(II) sulfate, 95.5% min

CAS: 7488-55-3 | O4SSn | 214.77 g/mol

$138.05 - $397.19

Chemical Identifiers

CAS 7488-55-3
Molecular Formula O4SSn
Molecular Weight (g/mol) 214.77
MDL Number MFCD00011246
InChI Key OBBXFSIWZVFYJR-UHFFFAOYSA-L
Synonym stannous sulfate, tin ii sulfate, tin 2+ sulfate, unii-0mfe10j96e, sulfuric acid, tin 2+ salt 1:1, stannous sulfate, crystal, stannous sulfate, solutions, tin, ion sn2+ sulfate, tin ii sulfate 250g
PubChem CID 62643
IUPAC Name tin(2+);sulfate
SMILES [Sn++].[O-]S([O-])(=O)=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA1153730
View Documents
Thermo Scientific Chemicals
01153730
250 g
Each for $138.05
Only null left
 
AA11537A1
View Documents
Thermo Scientific Chemicals
011537A1
1 kg
Each for $397.19
Only null left
 
Description

Description

It is used in tin plating baths and it is a precursor for tin metal.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7488-55-3
214.77
OBBXFSIWZVFYJR-UHFFFAOYSA-L
62643
[Sn++].[O-]S([O-])(=O)=O
O4SSn
MFCD00011246
stannous sulfate, tin ii sulfate, tin 2+ sulfate, unii-0mfe10j96e, sulfuric acid, tin 2+ salt 1:1, stannous sulfate, crystal, stannous sulfate, solutions, tin, ion sn2+ sulfate, tin ii sulfate 250g
tin(2+);sulfate

Specifications

378°C (decomposition)
Powder
95.5% min.
MFCD00011246
Soluble in water and dilute H2SO4.
[Sn++].[O-]S([O-])(=O)=O
214.77
214.77
Tin(II) sulfate
7488-55-3
250 g
O4SSn
stannous sulfate, tin ii sulfate, tin 2+ sulfate, unii-0mfe10j96e, sulfuric acid, tin 2+ salt 1:1, stannous sulfate, crystal, stannous sulfate, solutions, tin, ion sn2+ sulfate, tin ii sulfate 250g
OBBXFSIWZVFYJR-UHFFFAOYSA-L
tin(2+);sulfate
62643
4.15 g/mL
Videos
Safety and Handling

Safety and Handling

P260-P264b-P271-P272-P280-P302+P352-P304+P340-P305+P351+P338-P310-P312-P314-P333+P313-P362-P501c

H315-H317-H318-H332-H335-H373

EINECSNumber : 231-302-2

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only