missing translation for 'onlineSavingsMsg'
Learn More

Tin(II) sulfate, 95.5% min

Catalog No. AA1153730
Change view
Click to view available options
Quantity:
250 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1153730 250 g
AA11537A1 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1153730 Supplier Thermo Scientific Chemicals Supplier No. 01153730

CAS: 7488-55-3 | O4SSn | 214.77 g/mol

It is used in tin plating baths and it is a precursor for tin metal.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7488-55-3
Molecular Formula O4SSn
Molecular Weight (g/mol) 214.77
MDL Number MFCD00011246
InChI Key OBBXFSIWZVFYJR-UHFFFAOYSA-L
Synonym stannous sulfate, tin ii sulfate, tin 2+ sulfate, unii-0mfe10j96e, sulfuric acid, tin 2+ salt 1:1, stannous sulfate, crystal, stannous sulfate, solutions, tin, ion sn2+ sulfate, tin ii sulfate 250g
PubChem CID 62643
IUPAC Name tin(2+);sulfate
SMILES [Sn++].[O-]S([O-])(=O)=O

Specifications

Melting Point 378°C (decomposition)
Physical Form Powder
Quantity 250 g
Assay Percent Range 95.5% min.
Solubility Information Soluble in water and dilute H2SO4.
Formula Weight 214.77
Density 4.15 g/mL
Chemical Name or Material Tin(II) sulfate
Hazard Category H315-H317-H318-H332-H335-H373
Precautionary Statement P260-P264b-P271-P272-P280-P302+P352-P304+P340-P305+P351+P338-P310-P312-P314-P333+P313-P362-P501c
EINECSNumber 231-302-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only