missing translation for 'onlineSavingsMsg'
Learn More

Tin(II) phthalocyanine, Thermo Scientific Chemicals

Catalog No. p-7050868
Change view
Click to view available options
Quantity:
0.5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3960702 0.5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA3960702 Supplier Thermo Scientific Chemicals Supplier No. 03960702

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Tin(II) phthalocyanine is used as an organic light-emitting diode (OLED) material.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 15304-57-1
Molecular Formula C32H16N8Sn
Molecular Weight (g/mol) 631.25
MDL Number MFCD00058867
InChI Key LLVONELOQJAYBZ-UHFFFAOYSA-N
Synonym tin ii phthalocyanine, tin phthalocyanine, snpc, phthalocyanatotin cento

Specifications

Quantity 0.5 g
UN Number UN3146
Solubility Information Insoluble in water.
Formula Weight 631.43
Physical Form Powder
Chemical Name or Material Tin(II) phthalocyanine
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOTIN COMPOUNDS, SOLID, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only