Learn More
Tin(II) oxide, 99%
CAS: 21651-19-4 | OSn | 134.71 g/mol
Supplier: Thermo Scientific Chemicals A178890E
Description
Tin(II) oxide is used as reducing agent, soft abrasive, and in preparation of stannous salts. It is used in the manufacture of copper ruby glass, and for illumination with UV light.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsTin(II) oxide is used as reducing agent, soft abrasive, and in preparation of stannous salts. It is used in the manufacture of copper ruby glass, and for illumination with UV light.
Solubility
Insoluble in water and alcohol.
Notes
Incompatible with materials like strong oxidizing agents, strong acids, strong reducing agents and alkali metals.
Specifications
| Tin(II) oxide | |
| 21651-19-4 | |
| OSn | |
| MFCD00011243 | |
| stannous oxide, tin ii oxide, tin oxide sno, tin monoxide, tin oxide sn2o2, stannane, oxo, stannanone, stannicoxide, oxostannanylidene, oxo stannane | |
| QHGNHLZPVBIIPX-UHFFFAOYSA-N | |
| oxotin | |
| 88989 | |
| 99% | |
| 6.45 |
| 1080°C (decomposition) | |
| 2500 g | |
| SnO | |
| 14,8787 | |
| Insoluble in water and alcohol. | |
| O=[Sn] | |
| 134.71 | |
| 134.69 | |
| Odorless |
Chemical Identifiers
| 21651-19-4 | |
| 134.71 | |
| QHGNHLZPVBIIPX-UHFFFAOYSA-N | |
| 88989 | |
| O=[Sn] |
| OSn | |
| MFCD00011243 | |
| stannous oxide, tin ii oxide, tin oxide sno, tin monoxide, tin oxide sn2o2, stannane, oxo, stannanone, stannicoxide, oxostannanylidene, oxo stannane | |
| oxotin |
Safety and Handling
GHS H Statement
H302
Harmful if swallowed.
P260-P271-P304+P340-P312-P314-P501c
H335-H373
EINECSNumber : 244-499-5
RTECSNumber : XQ3700000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only